6-Bromo-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Thermo Scientific
Warning

H63527.MD H63527.03
H63527.MDEA 122 EUR
H63527.MD H63527.03
6-Bromo-1,2,3,4-tetrahydroisoquinoline 95%
6-Bromo-1,2,3,4-tetrahydroisoquinoline

Purity: 95% CAS No.: 226942-29-6 Molecular Formula: C9H10BrN Formula Weight: 212.09 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD07374370

Formula: C₉H₁₀BrN
MDL Number: MFCD07374370
CAS Number: 226942-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR