6-Bromoquinazoline-2,4(1H,3H)-dione 97%

Supplier: Thermo Scientific
Warning

H63547.03 H63547.06
H63547.03EA 87.9 EUR
H63547.03 H63547.06
6-Bromoquinazoline-2,4(1H,3H)-dione 97%
6-Bromoquinazoline-2,4(1H,3H)-dione

Purity: 97% CAS No.: 88145-89-5 Molecular Formula: C8H5BrN2O2 Formula Weight: 241.04 Physical Form: Solid Appearance: Off-white MDL No.: MFCD00462868

Formula: C₈H₅BrN₂O₂
MW: 241,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462868
CAS Number: 88145-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR