4-Nitro-1H-indole-3-carbaldehyde 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Nitroindole-3-carboxaldehyde
H63595.03
H63595.06
H63595.03EA
114
EUR
H63595.03
H63595.06
4-Nitro-1H-indole-3-carbaldehyde 97%
4-Nitro-1H-indole-3-carbaldehyde
Purity: 97%
CAS No.: 10553-11-4
Molecular Formula: C9H6N2O3
Formula Weight: 190.16
Physical Form: Solid
Appearance: Yellow
Melting Point: 189-193°
MDL No.: MFCD04973979
Formula:
C₉H₆N₂O₃ MW: 190,158 g/mol Melting Pt: 189…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973979 CAS Number: 10553-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...