N-(tert-Butoxycarbonyl)-D-threonine 95%
Supplier: Thermo Fisher Scientific
H63647.06
H63647.14
H63647.06EA
108
EUR
H63647.06
H63647.14
N-(tert-Butoxycarbonyl)-D-threonine 95%
N-(tert-Butoxycarbonyl)-D-threonine
Purity: 95%
CAS No.: 55674-67-4
Molecular Formula: C9H17NO5
Formula Weight: 219.23
Physical Form: Solid
Beilstein Reference: 2051734
MDL No.: MFCD00037807
Formula:
C₉H₁₇NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00037807 CAS Number: 55674-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...