5-Bromotryptamine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H63653.MD
H63653.03
H63653.06
H63653.MDEA
95.2
EUR
H63653.MD
H63653.03
H63653.06
5-Bromotryptamine 97%
5-Bromotryptamine
Purity: 97%
CAS No.: 3610-42-2
Molecular Formula: C10H11BrN2
Formula Weight: 239.11
Physical Form: Solid
MDL No.: MFCD00130169
Formula:
C₁₀H₁₁BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD00130169 CAS Number: 3610-42-2 EINECS: 222-779-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...