(R)-1-[α-(4-Chlorophenyl)benzyl]piperazine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Chlorobenzhydryl Piperazine, (R)-1-((4-Chlorophenyl)(phenyl)methyl)piperazine
H63722.MD
H63722.03
H63722.06
H63722.MDEA
0
EUR
H63722.MD
H63722.03
H63722.06
(R)-1-[α-(4-Chlorophenyl)benzyl]piperazine 97%
(R)-1-[alpha-(4-Chlorophenyl)benzyl]piperazine
Purity: 97%
CAS No.: 300543-56-0
Molecular Formula: C17H19ClN2
Formula Weight: 286.8
Physical Form: Solid
Appearance: White
MDL No.: MFCD11519277
Formula:
C₁₇H₁₉ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD11519277 CAS Number: 300543-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...