BippyPhos 97%
Supplier: Thermo Scientific
Synonyms:
5-(DI-T-BUTYLPHOSPHINO)-1',3',5'-TRIPHENYL-1,4'-BI-1H-PYRAZOLE, 5-(Di-tert-butylphosphino)-1', 3', 5'-triphenyl-1'H-[1,4']bipyrazole, 5-(di-tert-Butylphosphino)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole, 5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole
H63864.MD
H63864.03
H63864.MDEA
99.8
EUR
H63864.MD
H63864.03
BippyPhos 97%
BippyPhos
Purity: 97%
CAS No.: 894086-00-1
Molecular Formula: C32H35N4P
Formula Weight: 506.62
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 169-173°
MDL No.: MFCD09038440
Formula:
C₃₂H₃₅N₄P MW: 506,634 g/mol Melting Pt: 169…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD09038440 CAS Number: 894086-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...