1-(Methylsulphonyl)piperazine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1-Methanesulphonylpiperazine
H63870.06
H63870.14
H63870.22
H63870.06EA
54.9
EUR
H63870.06
H63870.14
H63870.22
1-(Methylsulphonyl)piperazine 97%
1-(Methylsulphonyl)piperazine
Purity: 97%
CAS No.: 55276-43-2
Molecular Formula: C5H12N2O2S
Formula Weight: 164.23
Physical Form: Solid
MDL No.: MFCD02153225
Formula:
C₅H₁₂N₂O₂S Storage Temperature: Ambient |
MDL Number:
MFCD02153225 CAS Number: 55276-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...