Z-Glu-OtBu 95%
Supplier: Thermo Scientific
Synonyms:
1-tert-Butyl N-Carbobenzoxy-L-glutamate, N-Cbz-L-glutamic acid 1-tert-Butyl ester, 1-tert-Butyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic acid 1-tert-Butyl ester, Z-Glu-OtBu, 1-tert-Butyl N-Benzyloxycarbonyl-L-glutamate, N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester, N-Benzyloxycarbonyl-L-glutamic acid 1-tert-Butyl ester
H63954.03
H63954.06
H63954.03EA
106
EUR
H63954.03
H63954.06
Z-Glu-OtBu 95%
Z-Glu-OtBu
Purity: 95%
CAS No.: 5891-45-2
Molecular Formula: C17H23NO6
Formula Weight: 337.37
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00077016
Formula:
C₁₇H₂₃NO₆ MW: 337,372 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00077016 CAS Number: 5891-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...