4,6-Dichloro-5-pyrimidinecarboxylic acid 95%

Supplier: Thermo Scientific
Warning

H63976.MD
H63976.MDEA 21.6 EUR
H63976.MD
4,6-Dichloro-5-pyrimidinecarboxylic acid 95%
4,6-Dichloro-5-pyrimidinecarboxylic acid

Purity: 95% CAS No.: 87600-98-4 Molecular Formula: C5H2Cl2N2O2 Formula Weight: 192.99 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Off-white MDL No.: MFCD09909611

Formula: C₅H₂Cl₂N₂O₂
MW: 192,99 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09909611
CAS Number: 87600-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR