7-Bromoquinoline 95%
Supplier: Thermo Scientific
|
Warning
|
H63987.03
H63987.06
H63987.03EA
169
EUR
H63987.03
H63987.06
7-Bromoquinoline 95%
7-Bromoquinoline
Purity: 95%
CAS No.: 4965-36-0
Molecular Formula: C9H6BrN
Formula Weight: 208.06
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD03695823
Formula:
C₉H₆BrN MW: 208,06 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03695823 CAS Number: 4965-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...