Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 2-((dimethylamino)methylene)-3-oxobutanoate, Ethyl 2-acetyl-3-(dimethylamino)acrylate
H64073.03
H64073.06
H64073.14
H64073.03EA
0
EUR
H64073.03
H64073.06
H64073.14
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate 95%
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate
Purity: 95%
CAS No.: 51145-57-4
Molecular Formula: C9H15NO3
Formula Weight: 185.22
Physical Form: Liquid
Appearance: Pale red
MDL No.: MFCD00087563
Formula:
C₉H₁₅NO₃ MW: 185,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00087563 CAS Number: 51145-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...