2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 98+%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,6-Dichloro-7-deazapurine
H64227.03
H64227.06
H64227.03EA
135
EUR
H64227.03
H64227.06
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 98+%
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
Purity: 98+%
CAS No.: 90213-66-4
Molecular Formula: C6H3Cl2N3
Formula Weight: 188.01
Physical Form: Solid
Melting Point: 247-250°
MDL No.: MFCD08059278
Formula:
C₆H₃Cl₂N₃ MW: 188,01 g/mol Melting Pt: 247…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD08059278 CAS Number: 90213-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...