5-Hydroxypicolinic acid 98+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-hydroxypyridine-2-carboxylic acid

H64265.03 H64265.06 H64265.14
H64265.03EA 36 EUR
H64265.03 H64265.06 H64265.14
5-Hydroxypicolinic acid 98+%
5-Hydroxypicolinic acid

Purity: 98+% CAS No.: 15069-92-8 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Physical Form: Solid Melting Point: 260-264° Beilstein Reference: 116009 MDL No.: MFCD00661309

Formula: C₆H₅NO₃
Melting Pt: 260…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00661309
CAS Number: 15069-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR