Methyl-2-amino-5-(trifluoromethyl)benzoate 98%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 2-amino-5-(trifluoromethyl)benzoate

H64283.03 H64283.06
H64283.03EA 108 EUR
H64283.03 H64283.06
Methyl-2-amino-5-(trifluoromethyl)benzoate 98%
Methyl-2-amino-5-(trifluoromethyl)benzoate

Purity: 98% CAS No.: 117324-58-0 Molecular Formula: C9H8F3NO2 Formula Weight: 219.16 Physical Form: Solid Appearance: White MDL No.: MFCD08234902

Formula: C₉H₈F₃NO₂
MW: 219,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234902
CAS Number: 117324-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR