2-Bromo-3-fluoroisonicotinic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Bromo-3-fluoropyridine-4-carboxylic acid

H64290.06
H64290.06EA 279 EUR
H64290.06
2-Bromo-3-fluoroisonicotinic acid 98%
2-Bromo-3-fluoroisonicotinic acid

Purity: 98% CAS No.: 1211530-89-0 Molecular Formula: C6H3BrFNO2 Formula Weight: 220.00 Physical Form: Solid MDL No.: MFCD13185796

Formula: C₆H₃BrFNO₂
MW: 219,997 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185796
CAS Number: 1211530-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR