2-Bromo-3-(trifluoromethyl)aniline 98%

Supplier: Thermo Fisher Scientific
Warning

H64309.03 H64309.06
H64309.03EA 0 EUR
H64309.03 H64309.06
2-Bromo-3-(trifluoromethyl)aniline 98%
2-Bromo-3-(trifluoromethyl)aniline

Purity: 98% CAS No.: 58458-10-9 Molecular Formula: C7H5BrF3N Formula Weight: 240.02 Physical Form: Solid Appearance: Off-white Beilstein Reference: 2835356 UN No.: UN2811 MDL No.: MFCD01656033

Formula: C₇H₅BrF₃N
Storage Temperature: Ambient
MDL Number: MFCD01656033
CAS Number: 58458-10-9
EINECS: 261-261-6
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR