4-Bromo-4'-iodobiphenyl 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Bromo-4'-iodo-1,1'-biphenyl, 1-Bromo-4-(4-iodophenyl)benzene
H64355.14
H64355.22
H64355.14EA
148
EUR
H64355.14
H64355.22
4-Bromo-4'-iodobiphenyl 98%
4-Bromo-4'-iodobiphenyl
Purity: 98%
CAS No.: 105946-82-5
Molecular Formula: C12H8BrI
Formula Weight: 359.00
Physical Form: Solid
Sensitivity: Light Sensitive
Melting Point: 175-179° lit.
Beilstein Reference: 2518914
UN No.: UN3152
MDL No.: MFCD17676204
Formula:
C₁₂H₈BrI MW: 359,00 g/mol Melting Pt: 175…179 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD17676204 CAS Number: 105946-82-5 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...