2-Bromo-5-chloropyridine 98%
Supplier: Thermo Scientific
|
Warning
|
H64367.14
H64367.22
H64367.14EA
76.5
EUR
H64367.14
H64367.22
2-Bromo-5-chloropyridine 98%
2-Bromo-5-chloropyridine
Purity: 98%
CAS No.: 40473-01-6
Molecular Formula: C5H3BrClN
Formula Weight: 192.44
Physical Form: Solid
Melting Point: 65-69° lit.
Boiling Point: 128°/16mm
Beilstein Reference: 109098
MDL No.: MFCD00234006
Formula:
C₅H₃BrClN Boiling Pt: 128 °C (16 mmHg) Melting Pt: 65…69 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00234006 CAS Number: 40473-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...