N-(Trimethylsilylmethyl)benzylamine 95%
Supplier: Thermo Scientific
|
Warning
|
H64450.14
H64450.14EA
0
EUR
H64450.14
N-(Trimethylsilylmethyl)benzylamine 95%
N-(Trimethylsilylmethyl)benzylamine
Purity: 95%
CAS No.: 53215-95-5
Molecular Formula: C11H19NSi
Formula Weight: 193.36
Flash Point: 96°(205°F)
Physical Form: Liquid
Density: 0.88
Refractive Index: 1.4960
Appearance: Pale yellow
Boiling Point: 89-90°/5mm
Beilstein Reference: 2802488
MDL No.: MFCD00012192
Formula:
C₁₁H₁₉NSi MW: 193,36 g/mol Boiling Pt: 89…90 °C (5 mmHg) Density: 0,88 Flash Pt: 96 °C (205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00012192 CAS Number: 53215-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...