6-Hydroxy-1,2,3,4-tetrahydroisoquinoline 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,2,3,4-Tetrahydroisoquinolin-6-ol
H64512.MD
H64512.03
H64512.MDEA
194
EUR
H64512.MD
H64512.03
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline 97%
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
Purity: 97%
CAS No.: 14446-24-3
Molecular Formula: C9H11NO
Formula Weight: 149.19
Physical Form: Solid
Melting Point: 192-193°
Beilstein Reference: 127578
MDL No.: MFCD01717047
Formula:
C₉H₁₁NO MW: 149,193 g/mol Melting Pt: 192…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD01717047 CAS Number: 14446-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...