2,3-Difluoro-6-methylpyridine 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,3-Difluoro-6-picoline
H64514.03
H64514.06
H64514.03EA
89.6
EUR
H64514.03
H64514.06
2,3-Difluoro-6-methylpyridine 98%
2,3-Difluoro-6-methylpyridine
Purity: 98%
Molecular Formula: C6H5F2N
Formula Weight: 129.11
Physical Form: Liquid
Appearance: Colourless to pale yellow
MDL No.: MFCD16610828
Formula:
C₆H₅F₂N MW: 129,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16610828 CAS Number: 1227579-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...