4-Bromo-9,9-dimethyl-9H-fluorene 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Bromo-9,9-dimethylfluorene
H64639.03
H64639.06
H64639.03EA
0
EUR
H64639.03
H64639.06
4-Bromo-9,9-dimethyl-9H-fluorene 98%
4-Bromo-9,9-dimethyl-9H-fluorene
Purity: 98%
CAS No.: 942615-32-9
Molecular Formula: C15H13Br
Formula Weight: 273.17
Physical Form: Solid
Beilstein Reference: 22540802
MDL No.: MFCD23099415
Formula:
C₁₅H₁₃Br MW: 273.173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23099415 CAS Number: 942615-32-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...