4-Chloro-6-methylpyrimidine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H64667.03
H64667.06
H64667.03EA
120
EUR
H64667.03
H64667.06
4-Chloro-6-methylpyrimidine 97%
4-Chloro-6-methylpyrimidine
Purity: 97%
CAS No.: 3435-25-4
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Physical Form: Solid
Appearance: Pale yellow
Beilstein Reference: 108656
MDL No.: MFCD02322991
Formula:
C₅H₅ClN₂ MW: 128.559 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02322991 CAS Number: 3435-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...