2-Amino-9,9-dimethylfluorene 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
9,9-Dimethyl-9H-fluoren-2-amine
H64771.03
H64771.06
H64771.03EA
0
EUR
H64771.03
H64771.06
2-Amino-9,9-dimethylfluorene 98%
2-Amino-9,9-dimethylfluorene
Purity: 98%
CAS No.: 108714-73-4
Molecular Formula: C15H15N
Formula Weight: 209.29
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 15506865
MDL No.: MFCD09953790
Formula:
C₁₅H₁₅N MW: 209,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09953790 CAS Number: 108714-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...