2-Aminopyridin-4-ol 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H64836.03
H64836.06
H64836.03EA
49.6
EUR
H64836.03
H64836.06
2-Aminopyridin-4-ol 98%
2-Aminopyridin-4-ol
Purity: 98%
CAS No.: 33631-05-9
Molecular Formula: C5H6N2O
Formula Weight: 110.12
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Pale yellow
Beilstein Reference: 385930
MDL No.: MFCD04114161
Formula:
C₅H₆N₂O |
MDL Number:
MFCD04114161 CAS Number: 33631-05-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...