3-Cyanophenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester

H64948.03 H64948.06
H64948.03EA 48.8 EUR
H64948.03 H64948.06
3-Cyanophenylboronic acid pinacol ester 97%
3-Cyanophenylboronic acid pinacol ester

Purity: 97% CAS No.: 214360-46-0 Molecular Formula: C13H16BNO2 Formula Weight: 229.09 Physical Form: Solid Appearance: White Melting Point: 78-82° dec. MDL No.: MFCD03789262

Formula: C₁₃H₁₆BNO₂
MW: 229,09 g/mol
Melting Pt: 78…82 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03789262
CAS Number: 214360-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR