Copper(I)-2-thiophenecarboxylate
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC
H64950.03
H64950.06
H64950.03EA
108
EUR
H64950.03
H64950.06
Copper(I)-2-thiophenecarboxylate
Copper(I)-2-thiophenecarboxylate
CAS No.: 68986-76-5
Molecular Formula: C5H3CuO2S
Formula Weight: 190.68
Physical Form: Powder
Sensitivity: Air and Light Sensitive
Appearance: Red to brown
MDL No.: MFCD02183524
Formula:
C₅H₃CuO₂S MW: 190,68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02183524 CAS Number: 68986-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...