5-(Carbobenzoxyamino)valeric acid 97%
Supplier: Thermo Scientific
Synonyms:
5-(Benzyloxycarbonylamino)valeric acid, 5-(Carbobenzoxyamino)pentanoic Acid, N-Cbz-5-aminovaleric Acid, 5-(((Benzyloxy)carbonyl)amino)pentanoic acid, N-Cbz-5-aminopentanoic Acid
H64958.03
H64958.06
H64958.03EA
41.5
EUR
H64958.03
H64958.06
5-(Carbobenzoxyamino)valeric acid 97%
5-(Carbobenzoxyamino)valeric acid
Purity: 97%
CAS No.: 23135-50-4
Molecular Formula: C13H17NO4
Formula Weight: 251.28
Physical Form: Solid
Appearance: Off-white
Melting Point: 99-105°
Beilstein Reference: 2334766
MDL No.: MFCD00188458
Formula:
C₁₃H₁₇NO₄ MW: 251,28 g/mol Melting Pt: 99…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00188458 CAS Number: 23135-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...