2-Amino-5-chlorophenol 97+%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Chloro-2-hydroxyaniline
H64978.14
H64978.22
H64978.14EA
87
EUR
H64978.14
H64978.22
2-Amino-5-chlorophenol 97+%
2-Amino-5-chlorophenol
Purity: 97+%
CAS No.: 28443-50-7
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Physical Form: Powder
Appearance: Brown
Melting Point: 145-153°
Beilstein Reference: 386421
UN No.: UN2811
MDL No.: MFCD02093863
Formula:
C₆H₆ClNO MW: 143,57 g/mol Melting Pt: 145…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093863 CAS Number: 28443-50-7 EINECS: 249-020-3 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...