(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate 95%

Supplier: Thermo Scientific
Warning

Synonyms: (S)-tert-Butyl 5-oxopyrrolidine-2-carboxylate, tert-Butyl L-Pyroglutamate, tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, tert-Butyl (S)-2-pyrrolidone-5-carboxylate

H65385.06 H65385.14
H65385.06EA 0 EUR
H65385.06 H65385.14
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate 95%
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate

Purity: 95% CAS No.: 35418-16-7 Molecular Formula: C9H15NO3 Formula Weight: 185.22 Physical Form: Powder Appearance: White Beilstein Reference: 3590226 MDL No.: MFCD06659481

Formula: C₉H₁₅NO₃
MW: 185,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659481
CAS Number: 35418-16-7
EINECS: 252-555-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR