3,5-Dimethylisoxazole-4-boronic acid pinacol ester 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole

H66042.06 H66042.14
H66042.06EA 107 EUR
H66042.06 H66042.14
3,5-Dimethylisoxazole-4-boronic acid pinacol ester 97%
3,5-Dimethylisoxazole-4-boronic acid pinacol ester

Purity: 97% CAS No.: 832114-00-8 Molecular Formula: C11H18BNO3 Formula Weight: 223.08 Physical Form: Solid Appearance: White to pale brown Melting Point: 95-99° MDL No.: MFCD05863910

Formula: C₁₁H₁₈BNO₃
MW: 223,08 g/mol
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD05863910
CAS Number: 832114-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR