1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene 97%
Supplier: Thermo Scientific
|
Danger
|
H66108.MD
H66108.03
H66108.MDEA
163
EUR
H66108.MD
H66108.03
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene 97%
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene
Purity: 97%
CAS No.: 3849-01-2
Molecular Formula: C12H15Cl3
Formula Weight: 265.60
Physical Form: Powder
Appearance: White
Beilstein Reference: 2369523
UN No.: UN3261
MDL No.: MFCD00229016
Formula:
C₁₂H₁₅Cl₃ Storage Temperature: Ambient |
MDL Number:
MFCD00229016 CAS Number: 3849-01-2 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...