N-Isopropylacrylamide 97%

Supplier: Thermo Fisher Scientific
Warning

H66262.14 H66262.22
H66262.14EA 0 EUR
H66262.14 H66262.22
N-Isopropylacrylamide 97%
N-Isopropylacrylamide

Purity: 97% CAS No.: 2210-25-5 Molecular Formula: C6H11NO Formula Weight: 113.16 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 60-63° Boiling Point: 89-92°/2mm Beilstein Reference: 1744926 MDL No.: MFCD00041913

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 89…92 °C (2 mmHg)
Melting Pt: 60…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041913
CAS Number: 2210-25-5
EINECS: 218-638-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR