2-Amino-2'-chloro-5-nitrobenzophenone 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Clonazepam Related Compound B, (2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone
H66440.06
H66440.14
H66440.22
H66440.06EA
24.5
EUR
H66440.06
H66440.14
H66440.22
2-Amino-2'-chloro-5-nitrobenzophenone 98%
2-Amino-2'-chloro-5-nitrobenzophenone
Purity: 98%
CAS No.: 2011-66-7
Molecular Formula: C13H9ClN2O3
Formula Weight: 276.68
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Pale yellow to yellow
Melting Point: 119-123°
Beilstein Reference: 2814216
MDL No.: MFCD00792453
Formula:
C₁₃H₉ClN₂O₃ MW: 276,68 g/mol Melting Pt: 119…123 °C |
MDL Number:
MFCD00792453 CAS Number: 2011-66-7 EINECS: 217-929-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...