Everolimus

Supplier: Thermo Scientific
Danger

Synonyms: (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,27-dihydroxy-3-((R)-1-((1S,3R,4R)-4-(2-hydroxyethoxy)-3-methoxycyclohexyl)propan-2-yl)-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-hexadecahydro-3H-23,27-epoxypyrido[2,1-c][1]oxa[4]azacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentaone

J60139.MA J60139.MB
J60139.MAEA 275 EUR
J60139.MA J60139.MB
Everolimus
Everolimus

CAS No.: 159351-69-6 Molecular Formula: C53H83NO14 Formula Weight: 958.22 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white Solubility: Soluble in DMSO or ethanol MDL No.: MFCD07785165


Applications: A rapamycin analog

Formula: C₅₃H₈₃NO₁₄
MW: 958,22 g/mol
MDL Number: MFCD00929329
CAS Number: 159351-69-6
Merck Index: 14,3907

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR