Docetaxel 99%
Supplier: Thermo Fisher Scientific
Synonyms:
Taxotere
J60174.MC
J60174.ME
J60174.03
J60174.MCEA
164
EUR
J60174.MC
J60174.ME
J60174.03
Docetaxel 99%
Docetaxel
Purity: 99%
CAS No.: 114977-28-5
Molecular Formula: C43H53NO14
Formula Weight: 807.88
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 186-192°
Solubility: Soluble in DMSO or ethanol
MDL No.: MFCD00800737
Applications: A second-generation cytotoxic antimicrotubule agent
Formula:
C₄₃H₅₃NO₁₄ MW: 807.89 g/mol Melting Pt: 186…192 °C |
MDL Number:
MFCD00800737 CAS Number: 114977-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...