Bortezomib 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
((R)-3-Methyl-1-((S)-3-phenyl-2-(pyrazine-2-carboxamido)propanamido)butyl)boronic acid
J60378.MA
J60378.MB
J60378.MAEA
305
EUR
J60378.MA
J60378.MB
Bortezomib 99%
Bortezomib
A potent, highly selective and reversible inhibitor of the 20S proteasome.
Applications: A potent, highly selective and reversible inhibitor of the 20S proteasome
Formula:
C₁₉H₂₅BN₄O₄ Melting Pt: 139…143 °C |
MDL Number:
MFCD09056737 CAS Number: 179324-69-7 ADR: 6.1,II Merck Index: 14,1351 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...