Diosgenin
Supplier: Thermo Fisher Scientific
CAS No.: 512-04-9
Molecular Formula: C27H42O3
Formula Weight: 414.63
Physical Form: Powder
Appearance: White
Melting Point: 200-206°
Beilstein Reference: 94582
MDL No.: MFCD00016887
Applications: Useful for synthesis of steroid products. Can be converted to pregnenolone and progesterone
Formula:
C₂₇H₄₂O₃ Melting Pt: 200…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016887 CAS Number: 512-04-9 EINECS: 208-134-3 Merck Index: 14,3295 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...