Dacarbazine
Supplier: Thermo Fisher Scientific
|
Danger
|
J61023.ME
J61023.03
J61023.MEEA
148
EUR
J61023.ME
J61023.03
Dacarbazine
Dacarbazine
CAS No.: 4342-03-4
Molecular Formula: C6H10N6O
Formula Weight: 182.18
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 210-212°
Solubility: Slightly soluble in water, DMSO and ethanol
MDL No.: MFCD00057167
Applications: Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA
Formula:
C₆H₁₀N₆O Melting Pt: 210…212 °C |
MDL Number:
MFCD00057167 CAS Number: 4342-03-4 EINECS: 224-396-1 Merck Index: 14,2798 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...