Tyr-Gly-Gly-Phe-Met

Supplier: Thermo Fisher Scientific

J61154.MB J61154.MF
J61154.MBEA 177 EUR
J61154.MB J61154.MF
Tyr-Gly-Gly-Phe-Met
Proteins and Peptides

Applications: Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors.


  • EINECS: 261-335-8


CAS No.: 58569-55-4 Molecular Formula: C27H35N5O7S Formula Weight: 573.67 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00076415


Applications: Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR