5-Iodo-2'-deoxycytidine 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidin-2(1H)-one
J61356.03
J61356.04
J61356.03EA
181
EUR
J61356.03
J61356.04
5-Iodo-2'-deoxycytidine 99%
5-Iodo-2'-deoxycytidine
Purity: 99%
CAS No.: 611-53-0
Molecular Formula: C9H12IN3O4
Formula Weight: 353.11
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 180-183°
MDL No.: MFCD00038063
Formula:
C₉H₁₂IN₃O₄ MW: 353.12 g/mol Melting Pt: 180…183 °C |
MDL Number:
MFCD00038063 CAS Number: 611-53-0 EINECS: 210-269-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...