Quisqualic acid ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 99+%
CAS No.: 52809-07-1
Molecular Formula: C5H7N3O5
Formula Weight: 189.13
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 185-187°
Beilstein Reference: 1078734
MDL No.: MFCD00069337
Applications: Group I mGlu receptor agonist. AMPA agonist
Formula:
C₅H₇N₃O₅ Melting Pt: 185…187 °C |
MDL Number:
MFCD00069337 CAS Number: 52809-07-1 Merck Index: 14,8085 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...