Tolnaftate
Supplier: Thermo Fisher Scientific
J61834.03
J61834.06
J61834.14
J61834.03EA
66
EUR
J61834.03
J61834.06
J61834.14
Tolnaftate
Tolnaftate
CAS No.: 2398-96-1
Molecular Formula: C19H17NOS
Formula Weight: 307.41
Physical Form: Powder
Appearance: White to off-white
Melting Point: 110-113°
Solubility: Soluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol
MDL No.: MFCD00056611
Applications: Antifungal agent
Formula:
C₁₉H₁₇NOS Melting Pt: 110…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00056611 CAS Number: 2398-96-1 EINECS: 219-266-6 Merck Index: 14,9519 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...