trans-Zeatin 97%
Supplier: Thermo Fisher Scientific
Synonyms:
6-(4-Hydroxy-3-methyl-2-butenylamino)purine
J61972.MF
J61972.MC
J61972.MD
J61972.MFEA
291
EUR
J61972.MF
J61972.MC
J61972.MD
trans-Zeatin 97%
trans-Zeatin
Purity: 97%
CAS No.: 1637-39-4
Molecular Formula: C10H13N5O
Formula Weight: 219.25
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
Melting Point: 208-210°
Solubility: Soluble in DMSO and DMF
Beilstein Reference: 616241
MDL No.: MFCD00213654
Applications: Plant growth hormone that stimulates cell division
Formula:
C₁₀H₁₃N₅O MW: 219.25 g/mol Melting Pt: 208…210 °C |
MDL Number:
MFCD00213654 CAS Number: 1637-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...