Bisacodyl ≥98%
Supplier: Thermo Fisher Scientific
Synonyms:
(Pyridin-2-ylmethylene)bis(4,1-phenylene) diacetate
J61997.09
J61997.14
J61997.09EA
133
EUR
J61997.09
J61997.14
Bisacodyl ≥98%
Bisacodyl
Purity: 98+%
CAS No.: 603-50-9
Molecular Formula: C22H19NO4
Formula Weight: 361.40
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White to off-white
MDL No.: MFCD00038039
Applications: A diphenolic laxative that stimulates adenyl cyclase activity
Formula:
C₂₂H₁₉NO₄ MW: 361.40 g/mol |
MDL Number:
MFCD00038039 CAS Number: 603-50-9 EINECS: 210-044-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...