Azathioprine

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 6-((1-Methyl-4-nitro-1H-imidazol-5-yl)thio)-9H-purine, 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

J62314.09 J62314.03 J62314.06
J62314.09EA 1010 EUR
J62314.09 J62314.03 J62314.06
Azathioprine
Azathioprine

CAS No.: 446-86-6 Molecular Formula: C9H7N7O2S Formula Weight: 277.26 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Appearance: Yellow MDL No.: MFCD00069203


Applications: An immunosuppressant. A purine synthesis inhibitor which inhibits the proliferation of cells

Formula: C₉H₇N₇O₂S
MW: 277.26 g/mol
MDL Number: MFCD00069203
CAS Number: 446-86-6
EINECS: 207-175-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR