Vinblastine sulphate 98%
Supplier: Thermo Scientific
|
Danger
|
J63598.MA
J63598.MF
J63598.MAEA
171
EUR
J63598.MA
J63598.MF
Vinblastine sulphate 98%
Vinblastine sulphate
Purity: 98%
CAS No.: 143-67-9
Molecular Formula: C46H60N4O13S
Formula Weight: 909.06
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 267°
Solubility: Soluble in water or DMSO
Beilstein Reference: 3659812
MDL No.: MFCD00082457
Applications: Anticancer agent; microtubule disrupter; Induces apoptosis
Formula:
C₄₆H₆₀N₄O₁₃S Melting Pt: 267 °C |
MDL Number:
MFCD00082457 CAS Number: 143-67-9 EINECS: 205-606-0 Merck Index: 14,9982 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...