2,2'-O-Cyclocytidine hydrochloride ≥98%
Supplier: Thermo Scientific
Synonyms:
Ancitabine Hydrochloride, Cyclocytidine hydrochloride, (2R,3R,3aS,9aR)-2-(Hydroxymethyl)-6-imino-3,3a,6,9a-tetrahydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-3-ol hydrochloride, 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride
J63845.03
J63845.06
J63845.03EA
56.5
EUR
J63845.03
J63845.06
2,2'-O-Cyclocytidine hydrochloride ≥98%
2,2'-O-Cyclocytidine hydrochloride
Purity: 98+%
CAS No.: 10212-25-6
Molecular Formula: C9H11N3O4.HCl
Formula Weight: 261.66
Physical Form: Powder
Appearance: White
Melting Point: 248-250°
MDL No.: MFCD00012636
Applications: Anti-tumor agent
Formula:
C₉H₁₁N₃O₄ MW: 261,66 g/mol Melting Pt: 248…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012636 CAS Number: 10212-25-6 EINECS: 233-515-6 Merck Index: 14,629 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...