4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid
Supplier: Thermo Scientific
Synonyms:
Tamibarotene
J64028.MA
J64028.MF
J64028.MAEA
162
EUR
J64028.MA
J64028.MF
4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid
4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid
CAS No.: 94497-51-5
Molecular Formula: C22H25NO3
Formula Weight: 351.44
Physical Form: Solid
Appearance: White
Solubility: Soluble to 50 mM in DMSO
MDL No.: MFCD00866188
Applications: A retinoic acid receptor (RAR) agonist
Formula:
C₂₂H₂₅NO₃ MW: 351,45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00866188 CAS Number: 94497-51-5 Merck Index: 14,9047 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...